Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfex™ Paddle Scraper with Handle

Catalog No. 17998560 Shop All Perfex Corp. Products
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Purple
Red
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-998-560 Yellow
17-998-554 Blue
17-998-555 Black
17-998-556 Green
17-998-557 Orange
17-998-558 Purple
17-998-559 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-998-560 Supplier Perfex™ Supplier No. 4640Y
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polymer industrial paddle scrapers with included handle, safe for food contact and ideal for scraping and spatula actions. Stiff and soft versions available for different applications.

  • Stiff polypropylene paddle scraper, ideal for stirring turbulence or scraping baked-on-food stuffs. Contoured blade with shapely corners for cleaning those hard-to-reach areas. Withstands heat up to 300°F .
  • Soft polyethylene paddle scraper, ideal for spatula applications. Contoured blade with shapely corners for cleaning those hard-to-reach areas. Withstands heat up to 150°F.
  • Impact-resistant, waterproof exterior is smooth and easy to clean. Comfort grip handle with hanging hole reduces hand strain and allows sanitary storage on tool hooks. Transfers maximum energy with minimum exertion to ensure small muscles are not overworked.
  • Stiff blades are available in eight colors to allow for color coding of your facility's subsections, eliminating possible cross-contamination. Available in White, Blue, Red, Green, Yellow, Orange, Purple and Black.
  • Soft blade is available in translucent white with white handle.

Specifications

Product Type Scraper
Material Polypropylene
Color Yellow
For Use With (Application) Mixing and scraping large batches of food material.
Includes Paddle scraper head and 34 in. handle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.