Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Paddle Sign - STOP/SLOW

Catalog No. 19169615 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Reflective Aluminum, Wood
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-169-615 Reflective Aluminum, Wood
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-169-615 Supplier Accuform™ Supplier No. FSP175
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accuform™ Paddle Signs help crossing guards and construction workers to direct and control traffic in school crossing zones or construction areas.

  • Message is printed on both sides
  • 18 in. reflective aluminum sign face
  • Includes 60 in. wood handle making the sign easy to carry and maneuver

Specifications

Includes 60 in. Wooden Handle
Material Reflective Aluminum, Wood
Mounting Type Floor-standing
For Use With (Application) Directing, and Controlling Traffic
Dimensions (L x W x H) 18 x 18 x 60 in.
Languages English
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.