Learn More
Ace Glass, Inc. Paddle stir shaft, stainless, 3 blade, 2.5in(63.5mm) diameter on 5/16(7.9mm) x 8.75in(222mm) shaft, high efficiency paddle for viscous liquids & semi-solids
SDP
Catalog No. 5097824
Encompass
Paddle stir shaft, stainless, 3 blade, 2.5in(63.5mm) diameter on 5/16(7.9mm) x 8.75in(222mm) shaft, high efficiency paddle for viscous liquids & semi-solids
Catalog No. 50-978-24 Supplier Ace Glass, Inc. Supplier No. 1364932
Paddle stir shaft, stainless, 3 blade, 2.5in(63.5mm) diameter on 5/16(7.9mm) x 8.75in(222mm) shaft, high efficiency paddle for viscous liquids & semi-solids