Learn More
Cole-Parmer Dynalon Polypropylene (PP) Paddled Stir Rod, 9-3/4" L; 10/Pk
Catalog No. NC0323982 Shop All Cole-Parmer Products
Encompass_Preferred
Use as a stir rod, policeman, or small spatula. Polypropylene (PP) rod features flat and triangular paddles at opposite ends. Max temperature is 266°F (130°C). Autoclavable.
Catalog No. NC0323982 Supplier Cole-Parmer Supplier No. 0637300
Use as a stir rod, policeman, or small spatula. Polypropylene (PP) rod features flat and triangular paddles at opposite ends. Max temperature is 266°F (130°C). Autoclavable.