Learn More
Microsolv Technology Corporation Paddles for sealing Film or Mats in the U-2D Micro-Sample Management System. 5/PK.
Catalog No. 502270500
Encompass_Preferred
Complete application of Sealing Films or Mats by pressing down on the Film or Mats with this Paddle. Assure that a secure uniform seal around all wells and inserts are complete by a gentle burnishing action. Use with the U-2D Brand Micro-Sample Management System or with other 96 Well Plates. The paddles fit within the rim of raised-well plates.
This item is not returnable. View return policy
Catalog No. 50-227-0500 Supplier Microsolv Technology Corporation Supplier No. 95296PDLPK
Complete application of Sealing Films or Mats by pressing down on the Film or Mats with this Paddle. Assure that a secure uniform seal around all wells and inserts are complete by a gentle burnishing action. Use with the U-2D Brand Micro-Sample Management System or with other 96 Well Plates. The paddles fit within the rim of raised-well plates.