Learn More
Med Vet International Cast Padding Undercast Webril™ 6 Inch X 4 Yard Cotton NonSterile, 3489, 6/Bag
Catalog No. 503043839
Encompass_Preferred
This nonsterile Webril undercast padding provides cushioning, moisture control and airflow for comfort It's suitable for use with either synthetic or plaster casting material The plaster cast roll will keep its consistency whether it's wet or dry Its flexible construction allows it to stretch while preventing it from moving around or bunching up The 6 inch x 4 yard roll of padding is made from 100% purified cotton fibers
Catalog No. 50-304-3839 Supplier Med Vet International Supplier No. 10064BG
This nonsterile Webril undercast padding provides cushioning, moisture control and airflow for comfort It's suitable for use with either synthetic or plaster casting material The plaster cast roll will keep its consistency whether it's wet or dry Its flexible construction allows it to stretch while preventing it from moving around or bunching up The 6 inch x 4 yard roll of padding is made from 100% purified cotton fibers