Learn More
Med Vet International Cast Padding Undercast Specialist 4 Inch X 4 Yard Cotton / Rayon NonSterile, 9044, 72/Case
Catalog No. 503046383
Encompass_Preferred
Conformability – the micropleated fibers fit every contour without slipping Cotton synthetic blend – the moisture from the casting material relaxes the crimped fibers resulting in a uniformly smooth surface Cotton/rayon blend is highly absorbent Strong enough to support, while still being tearable 10 centimeters x 3.6 meters
Catalog No. 50-304-6383 Supplier Med Vet International Supplier No. 4894CS
Conformability – the micropleated fibers fit every contour without slipping Cotton synthetic blend – the moisture from the casting material relaxes the crimped fibers resulting in a uniformly smooth surface Cotton/rayon blend is highly absorbent Strong enough to support, while still being tearable 10 centimeters x 3.6 meters