Learn More
Agilent Technologies PADEASY-1 VECTOR
Catalog No. 50125387
Encompass
The pAdEasy-1 vector is a 33.4 kb plasmid containing most of the human adenovirus serotype 5 (Ad5) genome, with deletions in the genes E1 and E3. This vector has been developed to infect but not replicate in non-permissive target cells
Catalog No. 50-125-387 Supplier Agilent Technologies Supplier No. 240005
The pAdEasy-1 vector is a 33.4 kb plasmid containing most of the human adenovirus serotype 5 (Ad5) genome, with deletions in the genes E1 and E3. This vector has been developed to infect but not replicate in non-permissive target cells