Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ PADI1 Recombinant Protein Antigen
SDP

Catalog No. NBP237844
Change view
Click to view available options
Quantity:
5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237844 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237844 Supplier Novus Biologicals™ Supplier No. NBP237844

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Specifications

Description Tissue was fixed in formalin immediately after excision, and embedded in paraffin
Quantity 5
Regulatory Status Research use only
Species Human
Specimen Type Tumor
Tissue Condition Malignant Melanoma
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.