Learn More
Aviva Systems Biology Corp PADI2 Antibody - middle region (ARP52328_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52328_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PADI2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PADI2. Alternate names: KIAA0994, PAD-H19, PAD2, PDI2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60713