Learn More
Medchemexpress LLC Peptidyl arginine deiminase 4, human (biotinylated, His-tagged) | >95.0% | 20UG
SDP
Catalog No. 5000177089
Encompass_Preferred
Recombinant full-length human peptidyl arginine deiminase 4 (PADI4), biotinylated and N-terminal His-tagged, supplied as a 20 μg solution formulated in 20 mM Tris, 500 mM NaCl, 0.5 mM TCEP, 1 mM EDTA, 10% glycerol, pH 7.6. Intended for biochemical assays, affinity capture, and binding studies using biotin-streptavidin or His-tag detection.
  • Full-length sequence (M1-P663)
  • Biotinylated for streptavidin-based capture and detection
  • N-terminal His tag for purification and detection
  • Greater than 95.0% purity by reducing SDS-PAGE
  • Molecular weight approximately 76.6 kDa
  • Expressed in Escherichia coli
  • Formulated in stabilizing buffer (Tris, NaCl, TCEP, EDTA, glycerol)
  • Supplied as 20 μg aliquot, recommended storage at -80 °C
Catalog No. 50-001-77089 Supplier Medchemexpress LLC Supplier No. HYP70103020UG
Recombinant full-length human peptidyl arginine deiminase 4 (PADI4), biotinylated and N-terminal His-tagged, supplied as a 20 μg solution formulated in 20 mM Tris, 500 mM NaCl, 0.5 mM TCEP, 1 mM EDTA, 10% glycerol, pH 7.6. Intended for biochemical assays, affinity capture, and binding studies using biotin-streptavidin or His-tag detection.
  • Full-length sequence (M1-P663)
  • Biotinylated for streptavidin-based capture and detection
  • N-terminal His tag for purification and detection
  • Greater than 95.0% purity by reducing SDS-PAGE
  • Molecular weight approximately 76.6 kDa
  • Expressed in Escherichia coli
  • Formulated in stabilizing buffer (Tris, NaCl, TCEP, EDTA, glycerol)
  • Supplied as 20 μg aliquot, recommended storage at -80 °C