Learn More
Hach Company Paddle Tester, Total Aerobic Bacteria/Yeast and Mold, 10/pk
Catalog No. NC2046511 Shop All Hach Company Products
Encompass
Hach Paddle Testers allow fast, easy screening of water samples, solid surfaces, and nonviscous liquids where unsanitary conditions may exist. The paddle is a double-sided slide attached to the vial cap. Each side of the slide is used to perform a separate test. The slide has a molded-in grid for easy colony counting without opening the optically clear, leakproof vial. Paddle Tester, Total Aerobic Bacteria/Yeast and Mold (Tryptone Glucose Extract Agar/Sabouraud Dextrose Agar), 10/pk.
This item is not returnable. View return policy
Catalog No. NC2046511 Supplier Hach Company Supplier No. 2610810
Hach Paddle Testers allow fast, easy screening of water samples, solid surfaces, and nonviscous liquids where unsanitary conditions may exist. The paddle is a double-sided slide attached to the vial cap. Each side of the slide is used to perform a separate test. The slide has a molded-in grid for easy colony counting without opening the optically clear, leakproof vial. Paddle Tester, Total Aerobic Bacteria/Yeast and Mold (Tryptone Glucose Extract Agar/Sabouraud Dextrose Agar), 10/pk.