Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Padoprazanum hydrochloride | 2940882-02-8 | 98.7% | 10 MG
SDP

Catalog No. 5000298561
Encompass_Preferred

Padoprazanum hydrochloride is the hydrochloride salt of padoprazan, a potassium-competitive acid blocker supplied as a research reagent for inhibition of gastric acid secretion. It is provided in small milligram-scale pack sizes for laboratory use.

  • High chemical purity (98.7%).
  • Molecular weight 441.90 g/mol.
  • Chemical formula C19H21ClFN3O4S.
  • Available in small mg pack sizes suitable for assay and in vitro studies.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-98561 Supplier Medchemexpress LLC Supplier No. HY400540C10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Padoprazanum hydrochloride is the hydrochloride salt of padoprazan, a potassium-competitive acid blocker supplied as a research reagent for inhibition of gastric acid secretion. It is provided in small milligram-scale pack sizes for laboratory use.

  • High chemical purity (98.7%).
  • Molecular weight 441.90 g/mol.
  • Chemical formula C19H21ClFN3O4S.
  • Available in small mg pack sizes suitable for assay and in vitro studies.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.