Learn More
Med Vet International Dynarex Combine Pads, 1/Pouch, Sterile, 8" X 7.5", 240/Case
Catalog No. 502830850
Encompass_Preferred
For when high absorbency is necessary for heavy draining wounds. Designed for use where high absorbency is needed for heavy draining wounds Outer cover constructed of soft non-woven material Fluffy inner fill absorbs and disperses fluids Sealed edges to prevent leakage Sterile pads packed in peel down pouches for convenient aseptic handling Pouches packed in open trays for easy access
Catalog No. 50-283-0850 Supplier Med Vet International Supplier No. 3502
 Restricted Item
Add to Cart
Add to Cart
For when high absorbency is necessary for heavy draining wounds. Designed for use where high absorbency is needed for heavy draining wounds Outer cover constructed of soft non-woven material Fluffy inner fill absorbs and disperses fluids Sealed edges to prevent leakage Sterile pads packed in peel down pouches for convenient aseptic handling Pouches packed in open trays for easy access