Learn More
Med Vet International Dynarex Non-Adherent Pads - Sterile, 3" X 8", 600/Case
Catalog No. 502830239
Encompass_Preferred
Dynarex DynaSafety Non-Adherent Pads feature a highly absorbent inner layer to help prevent leakage. These sterile, cotton/polyester pads are designed to effectively protect cuts, scrapes, and burns. These Pads are constructed to minimize adherence to the wound. Dynarex DynaSafety Non-Adherent Pads are sterile cotton/polyester pads designed to effectively protect cuts, scrapes, and burns without sticking to the wound. 
Catalog No. 50-283-0239 Supplier Med Vet International Supplier No. 3438
Dynarex DynaSafety Non-Adherent Pads feature a highly absorbent inner layer to help prevent leakage. These sterile, cotton/polyester pads are designed to effectively protect cuts, scrapes, and burns. These Pads are constructed to minimize adherence to the wound. Dynarex DynaSafety Non-Adherent Pads are sterile cotton/polyester pads designed to effectively protect cuts, scrapes, and burns without sticking to the wound. 