Learn More
MSC PRO-SAFE Sorbent Pads, Universal, 15 x 17 in, 100/Pk.
Catalog No. 502834768
Encompass
Sorbent pads designed for universal use, made from 3-Ply bonded polypropylene with SMS laminate. Each package contains 100 pads, measuring 15 in. wide and 17 in. long, capable of absorbing up to 30 gal. These pads are suitable for absorbing a wide range of fluids.
  • Universal use application
  • Polypropylene material
  • Three-ply, bonded, SMS laminate construction
  • Absorbs universal fluids
  • Medium absorbency weight
  • Perforated for easy tearing
Catalog No. 50-283-4768 Supplier MSC Supplier No. 75331108
Sorbent pads designed for universal use, made from 3-Ply bonded polypropylene with SMS laminate. Each package contains 100 pads, measuring 15 in. wide and 17 in. long, capable of absorbing up to 30 gal. These pads are suitable for absorbing a wide range of fluids.
  • Universal use application
  • Polypropylene material
  • Three-ply, bonded, SMS laminate construction
  • Absorbs universal fluids
  • Medium absorbency weight
  • Perforated for easy tearing