Learn More
MSC Brady SPC Sorbents Rag Rug Matting Roll, Universal, 55 gal
Catalog No. 502834877
Encompass
This matting roll is an eco-friendly and highly absorbent solution for managing spills in various industrial and maintenance applications. Constructed from recycled cotton and polyester fibers, it is designed for universal absorption of oil, water-based, and solvent-based fluids. It provides excellent footing while masking dirt and absorbed spills, offering a durable and affordable option for high-traffic areas.
  • Eco-friendly and highly absorbent.
  • Constructed from recycled cotton and polyester fibers.
  • Designed for universal absorption of oil, water-based, and solvent-based fluids.
  • Heavy absorbency weight is ideal for continuous leaks and larger spills.
  • Provides excellent footing while masking dirt and absorbed spills.
  • Contains 75% recycled content.
  • Dimensions of 36 in wide by 150 ft long.
  • Absorbency capacity of 55 gal.
Catalog No. 50-283-4877 Supplier MSC Supplier No. 06487276
This matting roll is an eco-friendly and highly absorbent solution for managing spills in various industrial and maintenance applications. Constructed from recycled cotton and polyester fibers, it is designed for universal absorption of oil, water-based, and solvent-based fluids. It provides excellent footing while masking dirt and absorbed spills, offering a durable and affordable option for high-traffic areas.
  • Eco-friendly and highly absorbent.
  • Constructed from recycled cotton and polyester fibers.
  • Designed for universal absorption of oil, water-based, and solvent-based fluids.
  • Heavy absorbency weight is ideal for continuous leaks and larger spills.
  • Provides excellent footing while masking dirt and absorbed spills.
  • Contains 75% recycled content.
  • Dimensions of 36 in wide by 150 ft long.
  • Absorbency capacity of 55 gal.