Learn More
MSC Brady SPC Sorbents Sorbent Pads, Oil Only, Polypropylene, 36 gal
Catalog No. 502835030
Encompass
Brady SPC Sorbent Pads are engineered for high-speed oil-only applications. Constructed from polypropylene, these pads offer a 36 gal absorption capacity, making them suitable for containing and cleaning up solvents, oil, fuel, and coolants. They feature a 1-ply construction, providing medium durability and minimal linting without a coverstock.
  • Designed for oil only applications
  • Made from polypropylene for effective absorption
  • Features a 36 gal capacity per package
  • 1-Ply construction with medium durability
  • Medium linting and no coverstock
  • Absorbs solvents, oil, fuel, and coolants
  • White color
Catalog No. 50-283-5030 Supplier MSC Supplier No. 00260612
Brady SPC Sorbent Pads are engineered for high-speed oil-only applications. Constructed from polypropylene, these pads offer a 36 gal absorption capacity, making them suitable for containing and cleaning up solvents, oil, fuel, and coolants. They feature a 1-ply construction, providing medium durability and minimal linting without a coverstock.
  • Designed for oil only applications
  • Made from polypropylene for effective absorption
  • Features a 36 gal capacity per package
  • 1-Ply construction with medium durability
  • Medium linting and no coverstock
  • Absorbs solvents, oil, fuel, and coolants
  • White color