Learn More
MSC Brady SPC Sorbents Sorbent Pads, Universal Use, 15 in Wide, 19 in Long, 30 gal
Catalog No. 502835052
Encompass
Brady SPC Sorbent Pads are designed for universal absorption of water-based, oil-based, and chemical-based fluids. Featuring a single-ply meltblown polypropylene construction with MAXX technology, these pads offer enhanced strength and absorption without added weight. Perforations allow users to reduce waste by using only what is needed, making them suitable for various industrial settings and maintenance areas.
  • Highly versatile
  • Single-ply construction
  • Medium durability
  • Medium linting
  • No coverstock
  • Absorbs water, solvents, oil, fuel, coolants, and chemicals
  • Perforated design to reduce waste
  • Enhanced strength and absorption with MAXX technology
  • Suitable for industrial settings, machine shops, refineries, paper mills, and transportation and maintenance areas
Catalog No. 50-283-5052 Supplier MSC Supplier No. 88882923
Brady SPC Sorbent Pads are designed for universal absorption of water-based, oil-based, and chemical-based fluids. Featuring a single-ply meltblown polypropylene construction with MAXX technology, these pads offer enhanced strength and absorption without added weight. Perforations allow users to reduce waste by using only what is needed, making them suitable for various industrial settings and maintenance areas.
  • Highly versatile
  • Single-ply construction
  • Medium durability
  • Medium linting
  • No coverstock
  • Absorbs water, solvents, oil, fuel, coolants, and chemicals
  • Perforated design to reduce waste
  • Enhanced strength and absorption with MAXX technology
  • Suitable for industrial settings, machine shops, refineries, paper mills, and transportation and maintenance areas