Learn More
MSC Brady SPC Sorbents Sorbent Pads, Oil Only, 19 in, 51 gal, Polypropylene
Catalog No. 502835129
Encompass
Brady SPC Sorbents ENV Oil Absorbents are designed for oil clean-up on land or water. Made with MAXX Technology, these highly economical, contractor-grade polypropylene pads absorb petroleum-based liquids and repel water, floating indefinitely even when saturated. This makes them suitable for outdoor and marine applications.
  • Made with MAXX Technology for superior absorbency and greater strength.
  • Lower disposal costs due to lighter weight.
  • Economical, contractor-grade for oil clean-up on land or water.
  • Absorbs petroleum-based liquids and repels water.
  • Single-ply construction.
  • Floats indefinitely even when saturated with oil.
  • Smooth construction maximizes surface area for efficient absorption.
  • Packaged in a green colored bag for streamlined absorbent identification.
Catalog No. 50-283-5129 Supplier MSC Supplier No. 67727479
Brady SPC Sorbents ENV Oil Absorbents are designed for oil clean-up on land or water. Made with MAXX Technology, these highly economical, contractor-grade polypropylene pads absorb petroleum-based liquids and repel water, floating indefinitely even when saturated. This makes them suitable for outdoor and marine applications.
  • Made with MAXX Technology for superior absorbency and greater strength.
  • Lower disposal costs due to lighter weight.
  • Economical, contractor-grade for oil clean-up on land or water.
  • Absorbs petroleum-based liquids and repels water.
  • Single-ply construction.
  • Floats indefinitely even when saturated with oil.
  • Smooth construction maximizes surface area for efficient absorption.
  • Packaged in a green colored bag for streamlined absorbent identification.