Learn More
MSC Brady SPC Sorbents Sorbent Pads, Oil Only, 15 x 19 in, 33 gal
Catalog No. 502835338
Encompass
These oil-only sorbent pads are designed for effective clean-up of petroleum-based liquids on land or water. Made from polypropylene with MAXX Technology, they offer superior absorbency and strength, while repelling water.
  • Made with MAXX Technology for superior absorbency and greater strength.
  • Weigh less, leading to lower disposal costs.
  • Highly economical, contractor grade for oil clean-up on land or water.
  • Absorbs petroleum-based liquids and repels water.
  • Economical single-ply construction.
  • Floats indefinitely even when saturated with oil, ideal for outdoor and marine applications.
  • Smooth construction maximizes surface area for efficient absorption.
  • Packaged in a green colored bag for streamlined absorbent identification.
Catalog No. 50-283-5338 Supplier MSC Supplier No. 00676866
These oil-only sorbent pads are designed for effective clean-up of petroleum-based liquids on land or water. Made from polypropylene with MAXX Technology, they offer superior absorbency and strength, while repelling water.
  • Made with MAXX Technology for superior absorbency and greater strength.
  • Weigh less, leading to lower disposal costs.
  • Highly economical, contractor grade for oil clean-up on land or water.
  • Absorbs petroleum-based liquids and repels water.
  • Economical single-ply construction.
  • Floats indefinitely even when saturated with oil, ideal for outdoor and marine applications.
  • Smooth construction maximizes surface area for efficient absorption.
  • Packaged in a green colored bag for streamlined absorbent identification.