Learn More
MSC PIG Absorbent Mat Pad, Heavyweight, 22 gal Capacity, Polypropylene, Perforated
Catalog No. 502835385
Encompass
This absorbent mat pad lasts 2x longer than ordinary mats, designed for fewer change-outs. It effectively absorbs a wide range of industrial liquids including oils, coolants, solvents, and water, providing a versatile solution for cleaning up various spills.
  • Lasts 2x longer than ordinary mats for fewer change-outs.
  • Made from multiple layers of 100% polypropylene, thermally bonded for strength.
  • Exclusive dimple pattern speeds liquid wicking for faster, easier cleanup.
  • Highly absorbent, fine-fiber construction leaves no residue.
  • Absorbs oils, coolants, solvents, and water.
  • Dark gray color hides grime, keeping the mat on the job longer.
  • Can be wrung out and incinerated after use.
  • Dispenser box allows for easy storage and protection from dirt.
  • Easy-tear perforations reduce waste.
  • Heavy-weight construction for durability and absorbency.
Catalog No. 50-283-5385 Supplier MSC Supplier No. 03236833
This absorbent mat pad lasts 2x longer than ordinary mats, designed for fewer change-outs. It effectively absorbs a wide range of industrial liquids including oils, coolants, solvents, and water, providing a versatile solution for cleaning up various spills.
  • Lasts 2x longer than ordinary mats for fewer change-outs.
  • Made from multiple layers of 100% polypropylene, thermally bonded for strength.
  • Exclusive dimple pattern speeds liquid wicking for faster, easier cleanup.
  • Highly absorbent, fine-fiber construction leaves no residue.
  • Absorbs oils, coolants, solvents, and water.
  • Dark gray color hides grime, keeping the mat on the job longer.
  • Can be wrung out and incinerated after use.
  • Dispenser box allows for easy storage and protection from dirt.
  • Easy-tear perforations reduce waste.
  • Heavy-weight construction for durability and absorbency.