Learn More
MSC 3M Sorbent Pad, Oil Only, 19 x 17 in, 100 Each
Catalog No. 502835482
Encompass
This high capacity, low profile sorbent pad picks up petroleum-based fluids and will not absorb water. It is suitable for routine use, cleaning up small spills, and wiping contaminated areas. The pads can also be used on a work surface to anticipate a spill or as a wipe. Each bale contains 100 pads.
  • For use in oil only sorbent applications.
  • Scrim on one side increases strength and helps eliminate linting.
  • Pads are conveniently sized and may be re-used.
  • May be used on a work surface in anticipation of a spill.
  • Ideal for use under docks, in shallow water, rocky shorelines, marshes, and beaches.
Catalog No. 50-283-5482 Supplier MSC Supplier No. 72451628
This high capacity, low profile sorbent pad picks up petroleum-based fluids and will not absorb water. It is suitable for routine use, cleaning up small spills, and wiping contaminated areas. The pads can also be used on a work surface to anticipate a spill or as a wipe. Each bale contains 100 pads.
  • For use in oil only sorbent applications.
  • Scrim on one side increases strength and helps eliminate linting.
  • Pads are conveniently sized and may be re-used.
  • May be used on a work surface in anticipation of a spill.
  • Ideal for use under docks, in shallow water, rocky shorelines, marshes, and beaches.