Learn More
MSC New Pig Absorbent Mat Roll, Lightweight Universal, Polypropylene, 30 in W x 150 ft L
Catalog No. 502834976
Encompass
This absorbent mat roll is designed for efficient cleanup of various industrial liquids. Constructed from multiple layers of thermally bonded polypropylene, its lightweight, universal design offers high absorbency and durability for everyday tasks and spills.
  • Lasts 2x longer than ordinary mats for fewer change-outs.
  • Eight layers of 100% polypropylene are thermally bonded.
  • Resists ripping, tearing, or fraying, even when saturated.
  • Dimple pattern speeds liquid wicking for faster cleanup.
  • Highly absorbent, fine-fiber construction leaves no residue.
  • Absorbs common industrial liquids like oils, water, solvents, and coolants.
  • Flame-resistant material melts when exposed to high heat.
  • Can be wrung out and incinerated for waste reduction or fuels blending.
  • Easy-tear perforations allow custom sizing to reduce waste.
Catalog No. 50-283-4976 Supplier MSC Supplier No. 31279896
This absorbent mat roll is designed for efficient cleanup of various industrial liquids. Constructed from multiple layers of thermally bonded polypropylene, its lightweight, universal design offers high absorbency and durability for everyday tasks and spills.
  • Lasts 2x longer than ordinary mats for fewer change-outs.
  • Eight layers of 100% polypropylene are thermally bonded.
  • Resists ripping, tearing, or fraying, even when saturated.
  • Dimple pattern speeds liquid wicking for faster cleanup.
  • Highly absorbent, fine-fiber construction leaves no residue.
  • Absorbs common industrial liquids like oils, water, solvents, and coolants.
  • Flame-resistant material melts when exposed to high heat.
  • Can be wrung out and incinerated for waste reduction or fuels blending.
  • Easy-tear perforations allow custom sizing to reduce waste.