Learn More
MSC Brady SPC Absorbents Absorbent Roll, Universal, Heavy Weight, 15 in x 150 ft, Polypropylene
Catalog No. 502835237
Encompass
This absorbent roll is designed for continuous leaks and larger spills in manufacturing plants and similar environments. It features a tough camouflage pattern that hides spills and a durable, universal 2-ply construction for absorbing water-based, petroleum-based, and non-aggressive chemical fluids. Perforations allow for cost control and reduced waste.
  • Universal absorbency for water-based, petroleum, and non-aggressive chemical fluids.
  • Heavy weight for continuous leaks and larger spills.
  • Durable 2-ply construction for maximum durability.
  • Low-linting, dimpled non-woven material.
  • Perforated design allows use of only necessary amount, reducing waste.
  • Gray camo pattern helps hide absorbed fluids.
  • Made from polypropylene.
  • Ideal as walk-on mats in manufacturing plants.
Catalog No. 50-283-5237 Supplier MSC Supplier No. 00922377
This absorbent roll is designed for continuous leaks and larger spills in manufacturing plants and similar environments. It features a tough camouflage pattern that hides spills and a durable, universal 2-ply construction for absorbing water-based, petroleum-based, and non-aggressive chemical fluids. Perforations allow for cost control and reduced waste.
  • Universal absorbency for water-based, petroleum, and non-aggressive chemical fluids.
  • Heavy weight for continuous leaks and larger spills.
  • Durable 2-ply construction for maximum durability.
  • Low-linting, dimpled non-woven material.
  • Perforated design allows use of only necessary amount, reducing waste.
  • Gray camo pattern helps hide absorbed fluids.
  • Made from polypropylene.
  • Ideal as walk-on mats in manufacturing plants.