Learn More
MSC New Pig Sorbent Pads, Oil Only, Heavy Weight, 20 x 15 in, Perforated
Catalog No. 502835253
Encompass
New Pig sorbent pads are designed for absorbing oil-based liquids, oil, and fuel. These heavy-weight, perforated pads are made from polypropylene and have a capacity of 22 gallons per package.
  • Made from polypropylene
  • Designed for oil-based liquids, oil, and fuel
  • Heavy absorbency weight
  • 20 inch long and 15 inch wide
  • Perforated for easy use
  • 22 gallon capacity per package
  • Manufactured in the United States
Catalog No. 50-283-5253 Supplier MSC Supplier No. 79936647
New Pig sorbent pads are designed for absorbing oil-based liquids, oil, and fuel. These heavy-weight, perforated pads are made from polypropylene and have a capacity of 22 gallons per package.
  • Made from polypropylene
  • Designed for oil-based liquids, oil, and fuel
  • Heavy absorbency weight
  • 20 inch long and 15 inch wide
  • Perforated for easy use
  • 22 gallon capacity per package
  • Manufactured in the United States