Learn More
MSC Brady Sorbent Pad, Universal, Polypropylene, 15 x 19 in, 20.5 gal
Catalog No. 502835022
Encompass
These absorbent pads are strong, durable, and low-linting, suitable for nearly every industrial setting. They are designed to pick up water-based, petroleum-based, and non-aggressive chemical fluids, making them ideal for most general maintenance needs.
  • Three-ply meltblown polypropylene construction for increased durability and reduced linting.
  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids.
  • Perforations provide cost control and reduce waste by allowing you to use only what is needed.
  • Strong, durable, and low linting.
Catalog No. 50-283-5022 Supplier MSC Supplier No. 06487888
These absorbent pads are strong, durable, and low-linting, suitable for nearly every industrial setting. They are designed to pick up water-based, petroleum-based, and non-aggressive chemical fluids, making them ideal for most general maintenance needs.
  • Three-ply meltblown polypropylene construction for increased durability and reduced linting.
  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids.
  • Perforations provide cost control and reduce waste by allowing you to use only what is needed.
  • Strong, durable, and low linting.