Learn More
MSC New Pig Sorbent Roll, Universal, Heavy Weight, 150 ft Long, 40.2 gal
Catalog No. 502835081
Encompass
This heavy-weight sorbent roll is designed for universal use, effectively absorbing a variety of fluids including water, solvents, oil, and coolants. Made from polypropylene, this durable roll provides an efficient solution for managing spills in various environments.
  • Heavy absorbency weight
  • Suitable for universal applications
  • Overall length of 150 ft
  • Width of 30 in
  • Capacity of 40.2 gal per package
  • Constructed from polypropylene material
  • Absorbs water, solvents, universal liquids, oil, and coolants
Catalog No. 50-283-5081 Supplier MSC Supplier No. 70760020
This heavy-weight sorbent roll is designed for universal use, effectively absorbing a variety of fluids including water, solvents, oil, and coolants. Made from polypropylene, this durable roll provides an efficient solution for managing spills in various environments.
  • Heavy absorbency weight
  • Suitable for universal applications
  • Overall length of 150 ft
  • Width of 30 in
  • Capacity of 40.2 gal per package
  • Constructed from polypropylene material
  • Absorbs water, solvents, universal liquids, oil, and coolants