Learn More
MSC Brady SPC Absorbents Sorbent Roll, Universal, Heavy Absorbency, Perforated, 3-Ply
Catalog No. 502835199
Encompass
The MRO Plus Perforated Absorbent Roll is designed for universal use in industrial settings. It effectively absorbs water-based, petroleum-based, and non-aggressive chemical fluids. Its 3-ply meltblown polypropylene construction provides durability and reduced linting, making it suitable for continuous leaks and larger spills.
  • Highly versatile, absorbs water-based, petroleum-based, and non-aggressive chemical fluids.
  • Heavy weight design for continuous leaks and larger spills, reducing material usage.
  • Perforations allow for cost control and waste reduction.
  • 3-ply meltblown polypropylene construction ensures increased durability and reduced linting.
  • Suitable for various industrial applications.
  • Strong, durable, and features medium linting with a dimpled texture.
Catalog No. 50-283-5199 Supplier MSC Supplier No. 75535815
The MRO Plus Perforated Absorbent Roll is designed for universal use in industrial settings. It effectively absorbs water-based, petroleum-based, and non-aggressive chemical fluids. Its 3-ply meltblown polypropylene construction provides durability and reduced linting, making it suitable for continuous leaks and larger spills.
  • Highly versatile, absorbs water-based, petroleum-based, and non-aggressive chemical fluids.
  • Heavy weight design for continuous leaks and larger spills, reducing material usage.
  • Perforations allow for cost control and waste reduction.
  • 3-ply meltblown polypropylene construction ensures increased durability and reduced linting.
  • Suitable for various industrial applications.
  • Strong, durable, and features medium linting with a dimpled texture.