Learn More
MSC PADS ROLLS MATS
Catalog No. 502834752
Encompass
The PIG Industrial Absorbent Adhesive-Backed Grippy Floor Mat provides a secure, absorbent solution for industrial workstations and machinery. Its adhesive backing prevents shifting and bunching, reducing slip and fall risks, while effectively absorbing a wide range of liquids.
  • Stays securely in place with proprietary adhesive backing
  • Easy peel-up for quick changeouts, enhancing productivity
  • Adheres to concrete, vinyl, linoleum, ceramic tile, quarry tile, and laminate
  • Tough, spunbond top layer with heavy-duty zigzag stitching for durability
  • Multi-layer fine-fiber polypropylene absorbs oils, coolants, solvents, water
  • Liquidproof barrier layer prevents absorbed liquids from soaking through
  • Dark color hides grime for a cleaner appearance
  • Meets NFPA33 - Static Dissipative & OSHA Standards, NFSI Certified
Catalog No. 50-283-4752 Supplier MSC Supplier No. 92580125
The PIG Industrial Absorbent Adhesive-Backed Grippy Floor Mat provides a secure, absorbent solution for industrial workstations and machinery. Its adhesive backing prevents shifting and bunching, reducing slip and fall risks, while effectively absorbing a wide range of liquids.
  • Stays securely in place with proprietary adhesive backing
  • Easy peel-up for quick changeouts, enhancing productivity
  • Adheres to concrete, vinyl, linoleum, ceramic tile, quarry tile, and laminate
  • Tough, spunbond top layer with heavy-duty zigzag stitching for durability
  • Multi-layer fine-fiber polypropylene absorbs oils, coolants, solvents, water
  • Liquidproof barrier layer prevents absorbed liquids from soaking through
  • Dark color hides grime for a cleaner appearance
  • Meets NFPA33 - Static Dissipative & OSHA Standards, NFSI Certified