Learn More
MSC PRO-SAFE Sorbent Roll, Oil Only, 30 in Wide, 150 ft Long, 48 gal Capacity
Catalog No. 502834908
Encompass
This sorbent roll is designed for absorbing oil and petroleum-based fluids. Made from polypropylene, it offers a 48 gallon absorption capacity, and is 30 inches wide and 150 feet long.
  • Sorbent roll
  • For oil and petroleum based fluids
  • 30 inches wide
  • 150 feet long
  • 48 gallon capacity
  • Polypropylene material
  • Absorbs oil
  • Medium absorbency
Catalog No. 50-283-4908 Supplier MSC Supplier No. 58617069
This sorbent roll is designed for absorbing oil and petroleum-based fluids. Made from polypropylene, it offers a 48 gallon absorption capacity, and is 30 inches wide and 150 feet long.
  • Sorbent roll
  • For oil and petroleum based fluids
  • 30 inches wide
  • 150 feet long
  • 48 gallon capacity
  • Polypropylene material
  • Absorbs oil
  • Medium absorbency