Learn More
MSC Brady Sorbent Pads, Universal, 15x19 in, 22 gal, 100 Pack
Catalog No. 502834599
Encompass
These sorbent pads are designed for general purpose applications, offering a balance of performance and price. They are suitable for cleaning up water-based, oil-based, and chemical-based fluids due to their universal absorption capabilities.
  • Universal absorption: effectively cleans up water-based, oil-based, and chemical-based fluids
  • Durable construction: made from single-ply meltblown polypropylene with MAXX technology for added strength and absorption without added weight
  • Perforations: allow users to tear off only the necessary amount, helping to reduce waste
  • Versatile application: can be used in various settings including industrial environments, machine shops, refineries, paper mills, and transportation and maintenance areas for quick and efficient spill cleanup
Catalog No. 50-283-4599 Supplier MSC Supplier No. 08894354
These sorbent pads are designed for general purpose applications, offering a balance of performance and price. They are suitable for cleaning up water-based, oil-based, and chemical-based fluids due to their universal absorption capabilities.
  • Universal absorption: effectively cleans up water-based, oil-based, and chemical-based fluids
  • Durable construction: made from single-ply meltblown polypropylene with MAXX technology for added strength and absorption without added weight
  • Perforations: allow users to tear off only the necessary amount, helping to reduce waste
  • Versatile application: can be used in various settings including industrial environments, machine shops, refineries, paper mills, and transportation and maintenance areas for quick and efficient spill cleanup