Learn More
MSC New Pig Sorbent Mat, Universal, Heavy, 100 Pieces
Catalog No. 502834687
Encompass
This sorbent mat is engineered for universal use, capable of absorbing oils, coolants, solvents, and water. It is constructed from multiple layers of 100% polypropylene that are thermally bonded for enhanced strength and features an exclusive dimple pattern to accelerate liquid wicking, facilitating faster and easier cleanup. Made from recycled content.
  • Multiple layers of 100% polypropylene for market-leading strength
  • Exclusive dimple pattern speeds wicking for faster cleanup
  • Heavy absorbency weight for demanding applications
  • Overall length of 20 in, width of 15 in
  • Capacity of 22 gal per package
  • Perforated for easy tearing and customized sizing
  • Thickness of 1 mil
  • Made from recycled content
Catalog No. 50-283-4687 Supplier MSC Supplier No. 89432694
This sorbent mat is engineered for universal use, capable of absorbing oils, coolants, solvents, and water. It is constructed from multiple layers of 100% polypropylene that are thermally bonded for enhanced strength and features an exclusive dimple pattern to accelerate liquid wicking, facilitating faster and easier cleanup. Made from recycled content.
  • Multiple layers of 100% polypropylene for market-leading strength
  • Exclusive dimple pattern speeds wicking for faster cleanup
  • Heavy absorbency weight for demanding applications
  • Overall length of 20 in, width of 15 in
  • Capacity of 22 gal per package
  • Perforated for easy tearing and customized sizing
  • Thickness of 1 mil
  • Made from recycled content