Learn More
Cayman Chemical Paederosidic Acid, Purity: ≥98%, CAS: 18842-98-3, 10 mg
SDP
Catalog No. 502909399
Encompass_Preferred
An iridoid glycoside; inhibits the proliferation of NCI H1650, HCC827, and A549 cells (IC50s = 40.68, 42.58, and 36.48 µg/ml, respectively); protects against PTZ- or maximal electroshock-induced seizures in mice from 10-40 mg/kg; increases duration of pentobarbital-induced sleep and decreases the latency to sleep onset in mice
Catalog No. 50-290-9399 Supplier Cayman Chemical Supplier No. 3536310 MG
An iridoid glycoside; inhibits the proliferation of NCI H1650, HCC827, and A549 cells (IC50s = 40.68, 42.58, and 36.48 µg/ml, respectively); protects against PTZ- or maximal electroshock-induced seizures in mice from 10-40 mg/kg; increases duration of pentobarbital-induced sleep and decreases the latency to sleep onset in mice