Learn More
Medchemexpress LLC Paederosidic acid | 18842-98-3 | MFCD28124366 | 10 MG
SDP
Catalog No. 5000419333
Encompass_Preferred
Paederosidic acid, isolated from P. scandens, is a chemical compound exhibiting anticancer and anti-inflammation activities. It primarily acts by inducing mitochondria-mediated apoptosis in lung cancer cells.
  • Inhibits lung cancer cells by inducing mitochondria-mediated apoptosis.
  • Shows effects on NCI-H1650, HCC827, and A549 cells.
  • Up-regulates caspase-3, caspase-8, caspase-9, Bid, Bax.
  • Down-regulates Bcl-2.
  • Stimulates Cyto-C release from mitochondria.
Catalog No. 50-004-19333 Supplier Medchemexpress LLC Supplier No. HYN699810MG
Paederosidic acid, isolated from P. scandens, is a chemical compound exhibiting anticancer and anti-inflammation activities. It primarily acts by inducing mitochondria-mediated apoptosis in lung cancer cells.
  • Inhibits lung cancer cells by inducing mitochondria-mediated apoptosis.
  • Shows effects on NCI-H1650, HCC827, and A549 cells.
  • Up-regulates caspase-3, caspase-8, caspase-9, Bid, Bax.
  • Down-regulates Bcl-2.
  • Stimulates Cyto-C release from mitochondria.