Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Paederosidic acid | 18842-98-3 | MFCD28124366 | 5 MG
SDP

Catalog No. 5000420998
Encompass_Preferred

Paederosidic acid is isolated from *P. scandens* and exhibits anticancer and anti-inflammation activities. It inhibits lung cancer cells by inducing mitochondria-mediated apoptosis.

  • Isolated from *Paederia scandens*
  • Exhibits anticancer and anti-inflammation activities
  • Inhibits lung cancer cells by inducing mitochondria-mediated apoptosis
  • Molecular weight: 464.44
  • Formula: C18H24O12S
  • Appearance: Solid
  • Color: White to light yellow
  • Purity: 99.90%
  • Relative density: 1.67 g/cm³
  • Stable under recommended storage conditions
  • Soluble in DMSO: 100 mg/mL (215.31 mM)

Catalog No. 50-004-20998 Supplier Medchemexpress LLC Supplier No. HYN69985MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Paederosidic acid is isolated from *P. scandens* and exhibits anticancer and anti-inflammation activities. It inhibits lung cancer cells by inducing mitochondria-mediated apoptosis.

  • Isolated from *Paederia scandens*
  • Exhibits anticancer and anti-inflammation activities
  • Inhibits lung cancer cells by inducing mitochondria-mediated apoptosis
  • Molecular weight: 464.44
  • Formula: C18H24O12S
  • Appearance: Solid
  • Color: White to light yellow
  • Purity: 99.90%
  • Relative density: 1.67 g/cm³
  • Stable under recommended storage conditions
  • Soluble in DMSO: 100 mg/mL (215.31 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.