Learn More
ATCC Streptococcus pneumoniae
Supplier: ATCC 6326
This strain of Streptococcus pneumoniae is typed as serotype 26 (Danish designation 6B). This bacterial culture can be used in respiratory disease research.