Learn More
ATCC Marinobacter; nauticus
Catalog No. 502380338
Encompass
Marinobacter nauticus strain SP.17 is a bacterial strain that was isolated from polluted seawater and sediments near a petroleum refinery in Gulf of Fos, Mediterranean coast, France. This strain has applications in the degradation of eicosane and hydrocarbons.
Catalog No. 50-238-0338 Supplier ATCC Supplier No. 49840
Marinobacter nauticus strain SP.17 is a bacterial strain that was isolated from polluted seawater and sediments near a petroleum refinery in Gulf of Fos, Mediterranean coast, France. This strain has applications in the degradation of eicosane and hydrocarbons.