Learn More
ATCC Paeniclostridium sordellii
Catalog No. 502384684
Encompass
Paeniclostridium sordellii strain 211 [NCIB 10717] is a bacterial type strain that can be used as a quality control strain for API, BBL, IDS, VITEK, and MicroScan [Reg TM] products.
Catalog No. 50-238-4684 Supplier ATCC Supplier No. 9714
Paeniclostridium sordellii strain 211 [NCIB 10717] is a bacterial type strain that can be used as a quality control strain for API, BBL, IDS, VITEK, and MicroScan [Reg TM] products.