Learn More
Cayman Chemical Paeoniflorin, Purity: ≥98%, CAS: 23180-57-6, 50 mg
SDP
Catalog No. 502983307
Encompass_Preferred
A monoterpene glycoside that has analgesic and anti-inflammatory actions in mice
Catalog No. 50-298-3307 Supplier Cayman Chemical Supplier No. 1986150 MG
A monoterpene glycoside that has analgesic and anti-inflammatory actions in mice