Learn More
Selleck Chemical LLC Paeonol S2339-200mg
SDP
Catalog No. 503109581
Encompass_Preferred
Paeonol (Peonol) a phenolic compound extracted from Chinese herbs Paeonia suffruticosa (moutan cortex) and Cynanchum paniculatum inhibits MAO with an IC50 of about 50 M
Catalog No. 50-310-9581 Supplier Selleck Chemical LLC Supplier No. S2339200MG
Paeonol (Peonol) a phenolic compound extracted from Chinese herbs Paeonia suffruticosa (moutan cortex) and Cynanchum paniculatum inhibits MAO with an IC50 of about 50 M