Learn More
Medchemexpress LLC HY-N0159 500mg Medchemexpress, Paeonol CAS:552-41-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0159500MG
Encompass_Preferred
Medchemexpress, HY-N0159 500mg Paeonol CAS:552-41-0 Paeonol is an active extraction from the root of Paeonia suffruticosa, Paeonol inhibits MAO-A and MAO-B with IC50 of 54.6 μM and 42.5 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2776