Learn More
Medchemexpress LLC Paeonol-d3 | 55712-78-2 | 10 MG
SDP
Catalog No. 5000295786
Encompass_Preferred
Paeonol-d3 is a deuterium-labeled analogue of paeonol used as a tracer and internal standard in drug development and pharmacokinetic and metabolic studies. It is supplied as a powder and is suitable for quantitative analyses where isotopic labeling is required.
  • Deuterium-labeled compound for tracer studies.
  • Useful as an internal standard in quantitative analyses.
  • Provided as a powder for convenient handling and storage.
  • Available in small research pack sizes, such as 10 mg.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-95786 Supplier Medchemexpress LLC Supplier No. HYN0159S10MG
Paeonol-d3 is a deuterium-labeled analogue of paeonol used as a tracer and internal standard in drug development and pharmacokinetic and metabolic studies. It is supplied as a powder and is suitable for quantitative analyses where isotopic labeling is required.
  • Deuterium-labeled compound for tracer studies.
  • Useful as an internal standard in quantitative analyses.
  • Provided as a powder for convenient handling and storage.
  • Available in small research pack sizes, such as 10 mg.
  • Stable under recommended storage conditions for long-term use.