Learn More
New England Biolabs, Inc. PaeR7I – 2000 units
SDP
Catalog No. 50812093
Encompass
A restriction endonuclease that recognizes the sequence C^TCGA_G.
Catalog No. 50-812-093 Supplier New England Biolabs, Inc. Supplier No. R0177S
A restriction endonuclease that recognizes the sequence C^TCGA_G.