Learn More
Cayman Chemical PAF Acetylhydrolase 2humn100ug
SDP
Catalog No. 502933816
Encompass_Preferred
Source: Active recombinant N-terminal His-tagged protein expressed in E. coli • Amino acids: 2-392 • MW: 46.1 kDa
Catalog No. 50-293-3816 Supplier Cayman Chemical Supplier No. 24292100 UG
Source: Active recombinant N-terminal His-tagged protein expressed in E. coli • Amino acids: 2-392 • MW: 46.1 kDa