Learn More
Cayman Chemical PAF Acetylhydrolase (human) Polyclonal Antibody, 1 ea
SDP
Catalog No. 502972658
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to the C-terminal region of human PAF-AH • Host: Rabbit • Species Reactivity: (+) Human (-) Mouse • Cross Reactivity: (+) PAF-AH • Applications: WB • MW = 45.4 kDa
Catalog No. 50-297-2658 Supplier Cayman Chemical Supplier No. 1606031 EA
Immunogen: Synthetic peptide corresponding to the C-terminal region of human PAF-AH • Host: Rabbit • Species Reactivity: (+) Human (-) Mouse • Cross Reactivity: (+) PAF-AH • Applications: WB • MW = 45.4 kDa