Learn More
Cayman Chemical PAF Acetylhydrolase Assay Kit, 96 Well
SDP
Catalog No. 502895989
Encompass_Preferred
PAF-AH catalyzes the hydrolysis of the potent biologically-active phospholipid PAF, generating inactive lyso-PAF. Cayman’s PAF-AH assay kit provides an accurate and convenient method for measurement of PAF-AH activity. The assay uses 2-thio PAF which serves as a substrate for PAF-AH. Upon hydrolysis of the acetyl thioester bond at the sn-2 position by PAF-AH, free thiols are detected using Ellman’s reagent.
Catalog No. 50-289-5989 Supplier Cayman Chemical Supplier No. 76090196 WELL
PAF-AH catalyzes the hydrolysis of the potent biologically-active phospholipid PAF, generating inactive lyso-PAF. Cayman’s PAF-AH assay kit provides an accurate and convenient method for measurement of PAF-AH activity. The assay uses 2-thio PAF which serves as a substrate for PAF-AH. Upon hydrolysis of the acetyl thioester bond at the sn-2 position by PAF-AH, free thiols are detected using Ellman’s reagent.