Learn More
Cayman Chemical PAF ACtylhydrolase Blockng
SDP
Catalog No. 502911914
Encompass_Preferred
Peptide Sequence: Human C-terminal amino acids 420-441 (TNINTTNQHIMLQNSSGIEKYN){2618} • To be used in conjunction with Cayman’s PAF-AH Polyclonal Antibody (Item No. 160603) to block protein-antibody complex formation during immunochemical analysis of PAF-AH.
Catalog No. 50-291-1914 Supplier Cayman Chemical Supplier No. 360603200 UG
Peptide Sequence: Human C-terminal amino acids 420-441 (TNINTTNQHIMLQNSSGIEKYN){2618} • To be used in conjunction with Cayman’s PAF-AH Polyclonal Antibody (Item No. 160603) to block protein-antibody complex formation during immunochemical analysis of PAF-AH.