Learn More
Enzo Life Sciences 1-O-Hexadecyl-2-O-acetyl-sn-glycero-3-phosphorylcholine (5mg). CAS: 74389-68-7
SDP
Supplier:  Enzo Life Sciences BMLL1000005
Encompass_Preferred
Platelet activating factor (PAF) with a C16 alkyl group. Initiates receptor-mediated responses in platelets and a variety of other cells. Rabbit platelet PAF receptor KD=0.1 nM. EC50 for stimulation of phosphoinositide metabolism in hippocampal cell line: 1.2-1.5 nM. Alternative name: PAF C-16. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol (>100 mg/ml). Soluble in water (>100mg/ml) after sonication for 1 minute at 40°C. Long Term Storage: -20°C.
Catalog No. 50-201-1129