Learn More
Cayman Chemical PAF C-16 Carboxylic Acid, Purity: ≥95%, CAS: 129879-41-0, 10 mg
SDP
Catalog No. 502969023
Encompass_Preferred
Modified with a carboxylic acid group terminating the C-16 alkyl chain, providing a convenient site for chemical crosslinking
Catalog No. 50-296-9023 Supplier Cayman Chemical Supplier No. 1539210 MG
Modified with a carboxylic acid group terminating the C-16 alkyl chain, providing a convenient site for chemical crosslinking